Potassium tetranitropalladate(II) (≥28,3% Pd)

Supplier: Thermo Scientific
Warning

10537.03 10537.06
10537.03EA 111 EUR
10537.03 10537.06
Potassium tetranitropalladate(II) (≥28,3% Pd)
Potassium tetranitropalladate (II)

CAS No.: 13844-89-8 Molecular Formula: K2Pd(NO2)4 Formula Weight: 368.63 Physical Form: Crystalline Melting Point: 310° dec. MDL No.: MFCD00049788

Formula: K₂N₄O₈Pd
MW: 368,63 g/mol
Melting Pt: 310 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00049788
CAS Number: 13844-89-8
EINECS: 237-568-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR