Dipotassium hexabromoiridate (≥25,4% Ir)

Supplier: Thermo Fisher Scientific
Warning

Synonyms: Potassium hexabromoiridate(IV)

12651.03 12651.06
12651.03EA 237 EUR
12651.03 12651.06
Dipotassium hexabromoiridate (≥25,4% Ir)
Dipotassium hexabromoiridate

CAS No.: 19121-78-9 Molecular Formula: K2IrBr6 Formula Weight: 749.86 Physical Form: Crystalline Sensitivity: Hygroscopic Appearance: Black Odour: Odourless MDL No.: MFCD00054221

Formula: Br₆IrK₂
MW: 749.84 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00054221
CAS Number: 19121-78-9
EINECS: 242-827-1

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR