Tetrapotassium diphosphate ≥95%

Supplier: Thermo Scientific
Warning

Synonyms: Tetrapotassium diphosphate, Potassium diphosphate, Potassium pyrophosphate, tetra-Potassium pyrophosphate, Potassium pyrophospate

13436.A1 13436.A7
13436.A1EA 61.6 EUR
13436.A1 13436.A7
Tetrapotassium diphosphate ≥95%
tetra-Potassium diphosphate

Purity: 95% CAS No.: 7320-34-5 Molecular Formula: K4P2O7 Formula Weight: 330.34 Physical Form: Powder Density: 2.33 Appearance: White Melting Point: 1090° Boiling Point: dec. Solubility: Soluble in water Merck Reference: 14,7663 MDL No.: MFCD00011393

Formula: K₄O₇P₂
MW: 330,34 g/mol
Boiling Pt: Dec.
Melting Pt: 1090 °C
Density: 2,33
Storage Temperature: Ambient
MDL Number: MFCD00011393
CAS Number: 7320-34-5
EINECS: 230-785-7
Merck Index: 14,7663

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR