1,3-Bis(trimethylsilyl)benzene

Supplier: Thermo Scientific

Synonyms: 1,3-Phenylenebis(trimethylsilane)

17542.03 17542.06
17542.03EA 0 EUR
17542.03 17542.06
1,3-Bis(trimethylsilyl)benzene
1,3-Bis(trimethylsilyl)benzene
Formula: C₁₂H₂₂Si₂
MW: 222,48 g/mol
CAS Number: 2060-89-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR