Chlorosulphuric acid ≥99%
Supplier: Thermo Scientific
|
Danger
|
87977.30
87977.A1
87977.30EA
93.4
EUR
87977.30
87977.A1
Chlorosulphuric acid ≥99%
Chlorosulphuric acid
Purity: 99%
CAS No.: 7790-94-5
Molecular Formula: HSO3Cl
Formula Weight: 116.52
Physical Form: Liquid
Density: 1.753
Refractive Index: 1.433
Sensitivity: Moisture Sensitive
Appearance: Colourless to pale yellow
Odour: Pungent
Boiling Point: 158°
Merck Reference: 14,2165
UN No.: 1754
MDL No.: MFCD00011523
Formula:
HClO₃S Boiling Pt: 158 °C Melting Pt: –80 °C Density: 1,753 Storage Temperature: Ambient |
MDL Number:
MFCD00011523 CAS Number: 7790-94-5 EINECS: 232-234-6 UN: 1754 ADR: 8,I Merck Index: 14,2165 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...