1-Methyl-2-thiourea ≥97%
Supplier: Thermo Scientific
|
Danger
|
A10437.14
A10437.22
A10437.36
A10437.14EA
35.6
EUR
A10437.14
A10437.22
A10437.36
1-Methyl-2-thiourea ≥97%
1-Methyl-2-thiourea
Purity: 98+%
CAS No.: 598-52-7
Molecular Formula: C2H6N2S
Formula Weight: 90.15
pH: 7
Melting Point: 119-124°
Beilstein Reference: 506162
UN No.: 2811
MDL No.: MFCD00004938
Formula:
C₂H₆N₂S MW: 90,15 g/mol Melting Pt: 119…124 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004938 CAS Number: 598-52-7 EINECS: 209-936-6 UN: 2811 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...