Ethyl cyanoacetate ≥98%
Supplier: Thermo Fisher Scientific
|
Warning
|
A11498.0E
A11498.36
A11498.0EEA
198
EUR
A11498.0E
A11498.36
Ethyl cyanoacetate ≥98%
Ethyl cyanoacetate
Purity: 98+%
CAS No.: 105-56-6
Molecular Formula: C5H7NO2
Formula Weight: 113.12
Flash Point: 109°(228°F)
Refractive Index: 1.4180
Odour: Weak
Melting Point: -23°
Boiling Point: 208-210°
Beilstein Reference: 605871
Merck Reference: 14,3786
MDL No.: MFCD00001940
Formula:
C₅H₇NO₂ MW: 113.12 g/mol Boiling Pt: 208…210 °C Melting Pt: –23 °C Density: 1.063 Flash Pt: 109 °C (228 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001940 CAS Number: 105-56-6 EINECS: 203-309-0 Merck Index: 13,03821 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...