1,3-Diacetoxybenzene ≥98%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
m-Phenylene di(acetate)
A11547.18
A11547.30
A11547.18EA
32.9
EUR
A11547.18
A11547.30
1,3-Diacetoxybenzene ≥98%
1,3-Diacetoxybenzene
Purity: 98%
CAS No.: 108-58-7
Molecular Formula: C10H10O4
Formula Weight: 194.19
Flash Point:
Formula:
C₁₀H₁₀O₄ MW: 194,19 g/mol Boiling Pt: 278 °C Melting Pt: 142…143 °C Density: 1,179 Flash Pt: >110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008701 CAS Number: 108-58-7 EINECS: 203-596-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...