2-Amino-4,6-dihydroxypyrimidine 98%
Supplier: Thermo Scientific
Synonyms:
2-Amino-6-hydroxy-1H-pyrimidin-4-one, 2-Amino-6-hydroxypyrimidin-4(3H)-one
A11813.14
A11813.22
A11813.14EA
54.7
EUR
A11813.14
A11813.22
2-Amino-4,6-dihydroxypyrimidine 98%
2-Amino-4,6-dihydroxypyrimidine
Purity: 98%
CAS No.: 56-09-7
Molecular Formula: C4H5N3O2
Formula Weight: 127.10
Melting Point:
Formula:
C₄H₅N₃O₂ MW: 127,1 g/mol Boiling Pt: 394 °C (1013 hPa) Melting Pt: >300 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006094 CAS Number: 56-09-7 EINECS: 200-256-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...