Dibenzothiophene ≥98%

Supplier: Thermo Scientific
Warning

Synonyms: Dibenzo[b,d]thiophene

A12288.14 A12288.22 A12288.36
A12288.14EA 43 EUR
A12288.14 A12288.22 A12288.36
Dibenzothiophene ≥98%
Dibenzothiophene

Purity: 98% CAS No.: 132-65-0 Molecular Formula: C12H8S Formula Weight: 184.25 Flash Point: 170°(338°F) Melting Point: 97-100° Boiling Point: 332-333° Beilstein Reference: 121101 UN No.: 3077 MDL No.: MFCD00004969

Formula: C₁₂H₈S
MW: 184,26 g/mol
Boiling Pt: 332…333 °C
Melting Pt: 97…100 °C
Density: 1,1295 g/cm³ (125 °C)
Flash Pt: 170 °C (338 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004969
CAS Number: 132-65-0
EINECS: 205-072-9
ADR: 9,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR