2-(Methylthio)aniline ≥98%
Supplier: Thermo Fisher Scientific
|
Danger
|
A12762.06
A12762.14
A12762.22
A12762.06EA
43.8
EUR
A12762.06
A12762.14
A12762.22
2-(Methylthio)aniline ≥98%
2-(Methylthio)aniline
Purity: 98%
CAS No.: 2987-53-3
Molecular Formula: C7H9NS
Formula Weight: 139.23
Flash Point:
Formula:
C₇H₉NS Boiling Pt: 233…234 °C Density: 1,111 Flash Pt: >110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00007708 CAS Number: 2987-53-3 EINECS: 221-055-9 UN: 2810 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...