Trityl bromide ≥98%
Supplier: Thermo Fisher Scientific
|
Danger
|
Synonyms:
Bromotriphenylmethane, Triphenylbromomethane, Triphenylmethyl bromide
A12864.09
A12864.18
A12864.36
A12864.09EA
35.6
EUR
A12864.09
A12864.18
A12864.36
Trityl bromide ≥98%
Trityl bromide
Purity: 98%
CAS No.: 596-43-0
Molecular Formula: BrC(C6H5)3
Formula Weight: 323.24
Melting Point: 152-156°
Boiling Point: 230°/15mm
UN No.: 1759
MDL No.: MFCD00000120
Formula:
(C₆H₅)₃CBr MW: 323.23 g/mol Boiling Pt: 230 °C (15 mmHg) Melting Pt: 152…156 °C Density: 1.55 Storage Temperature: Ambient |
MDL Number:
MFCD00000120 CAS Number: 596-43-0 EINECS: 209-884-4 UN: 1759 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...