Phthalaldehyde ≥98%
Supplier: Thermo Fisher Scientific
|
Danger
|
Synonyms:
1,2-Phthalic dicarboxaldehyde, Phthaldialdehyde, o-Phthalaldehyde, OPA, Phthaldicarboxaldehyde, Benzene-1,2-dicarboxaldehyde
A13299.18
A13299.09
A13299.30
A13299.18EA
267
EUR
A13299.18
A13299.09
A13299.30
Phthalaldehyde ≥98%
Phthalaldehyde
Purity: 98%
CAS No.: 643-79-8
Molecular Formula: C8H6O2
Formula Weight: 134.14
Flash Point: 132°(269°F)
Melting Point: 53-57°
Beilstein Reference: 878317
UN No.: 2923
MDL No.: MFCD00003335
Formula:
C₈H₆O₂ MW: 134.13 g/mol Boiling Pt: 266 Melting Pt: 53…57 °C Density: 1.13 g/cm³ (20 °C) Flash Pt: 132 °C (269 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00003335 CAS Number: 643-79-8 EINECS: 211-402-2 UN: 2923 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...