trans-2-Hexenoic acid 96%

Supplier: Thermo Scientific
Danger

Synonyms: trans-Hex-2-enoic acid

A13571.09 A13571.18 A13571.30
A13571.09EA 30 EUR
A13571.09 A13571.18 A13571.30
trans-2-Hexenoic acid 96%
trans-2-Hexenoic acid

Purity: 96% CAS No.: 13419-69-7 Molecular Formula: C6H10O2 Formula Weight: 114.14 Flash Point: 125°(257°F) Refractive Index: 1.4385 Melting Point: 30-35° Boiling Point: 217° Beilstein Reference: 1720443 UN No.: 3261 MDL No.: MFCD00002705

Formula: C₆H₁₀O₂
MW: 114,14 g/mol
Boiling Pt: 217 °C
Melting Pt: 30…35 °C
Density: 0,965
Flash Pt: 125 °C (257 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002705
CAS Number: 13419-69-7
EINECS: 236-528-5
UN: 3261
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR