(4-Formylphenoxy)acetic acid ≥98%

Supplier: Thermo Scientific
Warning

Synonyms: 2-(4-Formylphenoxy)acetic acid

A13960.09 A13960.18 A13960.30
A13960.09EA 62.6 EUR
A13960.09 A13960.18 A13960.30
(4-Formylphenoxy)acetic acid ≥98%
(4-Formylphenoxy)acetic acid

Purity: 98% CAS No.: 22042-71-3 Molecular Formula: C9H8O4 Formula Weight: 180.16 Melting Point: 193-198° Beilstein Reference: 779885

Formula: (C₆H₄(CHO))OCH₂COOH
MW: 180,16 g/mol
Boiling Pt: 333 °C (1013 hPa)
Melting Pt: 193…198 °C
Storage Temperature: Ambient
MDL Number: MFCD00016613
CAS Number: 22042-71-3
EINECS: 244-749-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR