(Chloromethyl)trimethylsilane ≥98%

Supplier: Thermo Scientific
Danger

A14303.09 A14303.18 A14303.30
A14303.09EA 41.2 EUR
A14303.09 A14303.18 A14303.30
(Chloromethyl)trimethylsilane ≥98%
(Chloromethyl)trimethylsilane

Purity: 98% CAS No.: 2344-80-1 Molecular Formula: ClCH2Si(CH3)3 Formula Weight: 122.67 Flash Point: -2°(28°F) Refractive Index: 1.4180 Odour: Odourless Boiling Point: 97-98° Beilstein Reference: 906705 UN No.: 1993 MDL No.: MFCD00000878

Formula: ClH₂CSi(CH₃)₃
MW: 122,67 g/mol
Boiling Pt: 97…98 °C
Density: 0,886
Flash Pt: –2 °C (28 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000878
CAS Number: 2344-80-1
EINECS: 219-058-5
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR