Benzoylacetonitrile ≥98%

Supplier: Thermo Scientific
Danger

Synonyms: 3-Phenyl-3-oxopropanenitrile, 3-Oxo-3-phenylpropanenitrile

A14473.14 A14473.06 A14473.22
A14473.14EA 283 EUR
A14473.14 A14473.06 A14473.22
Benzoylacetonitrile ≥98%
Benzoylacetonitrile

Purity: 98+% CAS No.: 614-16-4 Molecular Formula: C9H7NO Formula Weight: 145.16 Melting Point: 79-83° Boiling Point: 159-160°/10mm MDL No.: MFCD00001942

Formula: (C₆H₅)C(O)CH₂CN
MW: 145,16 g/mol
Boiling Pt: 159…160 °C (10 mmHg)
Melting Pt: 79…83 °C
Storage Temperature: Ambient
MDL Number: MFCD00001942
CAS Number: 614-16-4
EINECS: 210-369-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR