α,α,α-Trifluoro-p-toluonitrile ≥98%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
4-(Trifluoromethyl)benzonitrile, 4-Cyanobenzotrifluoride, p-(Trifluoromethyl)benzonitrile
A14514.09
A14514.18
A14514.30
A14514.09EA
50.8
EUR
A14514.09
A14514.18
A14514.30
α,α,α-Trifluoro-p-toluonitrile ≥98%
α,α,α-Trifluoro-p-toluonitrile
Purity: 98%
CAS No.: 455-18-5
Molecular Formula: C8H4F3N
Formula Weight: 171.12
Flash Point: 71°(159°F)
Refractive Index: 1.4583
Melting Point: 36-40°
Boiling Point: 183-185°
Beilstein Reference: 2046478
UN No.: 3439
MDL No.: MFCD00001826
Formula:
C₈H₄F₃N MW: 171,12 g/mol Boiling Pt: 183…185 °C Melting Pt: 36…40 °C Density: 1,278 Flash Pt: 71 °C (159 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001826 CAS Number: 455-18-5 EINECS: 207-239-1 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...