2-Bromo-5-nitrothiazole 98

Supplier: Thermo Fisher Scientific
Warning

A14701.06 A14701.14
A14701.06EA 83.4 EUR
A14701.06 A14701.14
2-Bromo-5-nitrothiazole 98
2-Bromo-5-nitrothiazole

Purity: 98% CAS No.: 3034-48-8 Molecular Formula: C3HBrN2O2S Formula Weight: 209.03 Melting Point: 85-90° Beilstein Reference: 4856

Formula: C₃HBrN₂O₂S
MW: 209.02 g/mol
Melting Pt: 85…90 °C
Density: 2.295 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00005317
CAS Number: 3034-48-8
EINECS: 221-226-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR