5-Bromouracil ≥98%

Supplier: Thermo Scientific
Warning

Synonyms: 5-Bromopyrimidine-2,4(1H,3H)-dione, Bromouracil

A14799.06 A14799.14 A14799.22
A14799.06EA 31.4 EUR
A14799.06 A14799.14 A14799.22
5-Bromouracil ≥98%
5-Bromouracil

Purity: 98+% CAS No.: 51-20-7 Molecular Formula: C4H3BrN2O2 Formula Weight: 190.99 Melting Point: ca 310° dec. Beilstein Reference: 127176 Merck Reference: 14,1440 MDL No.: MFCD00006017

Formula: C₄H₃BrN₂O₂
MW: 190,98 g/mol
Boiling Pt: 384 °C (1013 hPa)
Melting Pt: ∼ 310 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00006017
CAS Number: 51-20-7
EINECS: 200-084-0
Merck Index: 14,1440

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR