2-Pyridylmethanol ≥98%

Supplier: Thermo Fisher Scientific
Warning

A15005.14 A15005.22 A15005.36
A15005.14EA 41.7 EUR
A15005.14 A15005.22 A15005.36
2-Pyridylmethanol ≥98%
2-Pyridylmethanol

Purity: 98+% CAS No.: 586-98-1 Molecular Formula: C6H7NO Formula Weight: 109.13 Storage Temperature: 2°C to 8°C Flash Point: 120°(248°F) Refractive Index: 1.5440 Melting Point: -6 to -5° Boiling Point: 112-113°/16mm Beilstein Reference: 107849 MDL No.: MFCD00006348

Formula: C₆H₇NO
Boiling Pt: 112…113 °C (16 mmHg)
Melting Pt: –6 to –5 °C
Density: 1,131
Flash Pt: 120 °C (248 °F)
MDL Number: MFCD00006348
CAS Number: 586-98-1
EINECS: 209-592-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR