(Iodomethyl)trimethylsilane ≥99%
Supplier: Thermo Fisher Scientific
|
Warning
|
A15120.09
A15120.18
A15120.09EA
89.4
EUR
A15120.09
A15120.18
(Iodomethyl)trimethylsilane ≥99%
(Iodomethyl)trimethylsilane
Purity: 99%
CAS No.: 4206-67-1
Molecular Formula: C4H11ISi
Formula Weight: 214.12
Flash Point: 31°(88°F)
Refractive Index: 1.4915
Odour: Recognizable
Boiling Point: 140-142°
Beilstein Reference: 1731485
UN No.: 1993
MDL No.: MFCD00001077
Formula:
C₄H₁₁ISi Boiling Pt: 140…142 °C Density: 1,443 Flash Pt: 31 °C (88 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001077 CAS Number: 4206-67-1 EINECS: 224-123-6 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...