9-Anthracenemethanol ≥97%

Supplier: Thermo Fisher Scientific

Synonyms: 9-Anthrylmethanol, Anthracene-9-methanol

A15188.09 A15188.18 A15188.30
A15188.09EA 99.5 EUR
A15188.09 A15188.18 A15188.30
9-Anthracenemethanol ≥97%
9-Anthracenemethanol

Purity: 97% CAS No.: 1468-95-7 Molecular Formula: C15H12O Formula Weight: 208.26 Melting Point: 160-165° Beilstein Reference: 1873402

Formula: C₁₅H₁₂O
Boiling Pt: 371 °C (1013 hPa)
Melting Pt: 160…165 °C
Density: 1,3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00001264
CAS Number: 1468-95-7
EINECS: 215-998-5
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR