N,N,N-Trimethylanilinium tribromide ≥97%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Phenyltrimethylammonium tribromide, Mono(N,N,N-trimethylbenzenaminium) tribromide, Trimethylphenylammonium tribromide

A15326.0B A15326.18 A15326.30
A15326.0BEA 889 EUR
A15326.0B A15326.18 A15326.30
N,N,N-Trimethylanilinium tribromide ≥97%
N,N,N-Trimethylanilinium tribromide

Purity: 97% CAS No.: 4207-56-1 Molecular Formula: C6H5N(CH3)3Br.Br2 Formula Weight: 375.93 Odour: Irritating Melting Point: 112-117° Beilstein Reference: 3919459 UN No.: 3261 MDL No.: MFCD00011789

Formula: C₉H₁₄Br₃N
MW: 375.93 g/mol
Melting Pt: 112…117 °C
Storage Temperature: Ambient
MDL Number: MFCD00011789
CAS Number: 4207-56-1
EINECS: 224-127-8
UN: 3261
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR