Indole-3-carbaldehyd ≥99%

Supplier: Thermo Fisher Scientific

A15330.14 A15330.22
A15330.14EA 81.5 EUR
A15330.14 A15330.22
Indole-3-carbaldehyd ≥99%
Indole-3-carbaldehyd

Purity: 99% CAS No.: 487-89-8 Molecular Formula: C9H7NO Formula Weight: 145.16 Flash Point: 240°(464°F) Melting Point: 195-198° Beilstein Reference: 114117 MDL No.: MFCD00005622

Formula: C₉H₇NO
Melting Pt: 195…198 °C
Density: 1,34 g/cm³ (20 °C)
Flash Pt: 240 °C (464 °F)
Storage Temperature: Ambient
MDL Number: MFCD00005622
CAS Number: 487-89-8
EINECS: 207-665-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR