(3,4-Xylyloxy)acetic acid ≥98%
Supplier: Thermo Fisher Scientific
|
Warning
|
A15355.06
A15355.14
A15355.06EA
0
EUR
A15355.06
A15355.14
(3,4-Xylyloxy)acetic acid ≥98%
(3,4-Xylyloxy)acetic acid
Purity: 98%
CAS No.: 13335-73-4
Molecular Formula: C10H12O3
Formula Weight: 180.20
Melting Point: 160-165°
Beilstein Reference: 2558866
Formula:
(C₆H₃(CH₃)₂)OCH₂COOH Melting Pt: 160…165 °C Storage Temperature: Ambient |
MDL Number:
MFCD00017595 CAS Number: 13335-73-4 EINECS: 236-386-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...