Tripropyl orthoformate ≥97%

Supplier: Thermo Scientific
Warning

Synonyms: 1,1',1''-[Methylidynetris(oxy)]trispropane, Tri-n-propyl orthoformate, Tripropylorthoformate

A15503.06 A15503.14 A15503.22
A15503.06EA 22.9 EUR
A15503.06 A15503.14 A15503.22
Tripropyl orthoformate ≥97%
Tripropyl orthoformate

Purity: 97% CAS No.: 621-76-1 Molecular Formula: HC(OCH2CH2CH3)3 Formula Weight: 190.28 Flash Point: 72°(161°F) Refractive Index: 1.4070 Boiling Point: 106-108°/40mm Beilstein Reference: 1744249 MDL No.: MFCD00015214

Formula: C₁₀H₂₂O₃
MW: 190,28 g/mol
Boiling Pt: 106…108 °C (40 mmHg)
Density: 0,886
Flash Pt: 72 °C (161 °F)
Storage Temperature: Ambient
MDL Number: MFCD00015214
CAS Number: 621-76-1
EINECS: 210-704-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR