4-Phenylbenzophenone ≥99%

Supplier: Thermo Fisher Scientific
Warning

A15573.14 A15573.22
A15573.14EA 60 EUR
A15573.14 A15573.22
4-Phenylbenzophenone ≥99%
4-Phenylbenzophenone

Purity: 99% CAS No.: 2128-93-0 Molecular Formula: C19H14O Formula Weight: 258.32 Melting Point: 98-103° Beilstein Reference: 1876092

Formula: C₁₉H₁₄O
Boiling Pt: 419 °C (1013 hPa)
Melting Pt: 98…103 °C
Density: 1,262 g/cm³ (0 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003079
CAS Number: 2128-93-0
EINECS: 218-345-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR