Crotonic acid 98%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
trans-Crotonic acid, 2-Butenoic acid, trans-2-Butenoic acid
A15765.0E
A15765.14
A15765.36
A15765.0EEA
304
EUR
A15765.0E
A15765.14
A15765.36
Crotonic acid 98%
Crotonic acid
Purity: 98%
CAS No.: 107-93-7
Molecular Formula: C4H6O2
Formula Weight: 86.09
Flash Point: 87°(188°F)
Melting Point: 70-74°
Boiling Point: 128°/100mm
Beilstein Reference: 1719943
Merck Reference: 14,2597
UN No.: 2823
MDL No.: MFCD00002701
Formula:
C₄H₆O₂ MW: 86,09 g/mol Boiling Pt: 128 °C (100 mmHg) Melting Pt: 70…74 °C Density: 1,027 Flash Pt: 87 °C (188 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002701 CAS Number: 107-93-7 EINECS: 203-533-9 UN: 2823 ADR: 8,III Merck Index: 14,2597 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...