3-Nitrophthalic anhydride ≥97%
Supplier: Thermo Scientific
|
Warning
|
A15857.14
A15857.22
A15857.14EA
89.7
EUR
A15857.14
A15857.22
3-Nitrophthalic anhydride ≥97%
3-Nitrophthalic anhydride
Purity: 97%
CAS No.: 641-70-3
Molecular Formula: C8H3NO5
Formula Weight: 193.11
Melting Point: 161-166°
Beilstein Reference: 179963
Formula:
C₈H₃NO₅ MW: 193,12 g/mol Melting Pt: 161…166 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005921 CAS Number: 641-70-3 EINECS: 211-373-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...