2-Amino-4-methylpyrimidine ≥98%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 4-Methylpyrimidin-2-ylamine, 4-Methylpyrimidin-2-amine

A16081.09 A16081.18
A16081.09EA 39.5 EUR
A16081.09 A16081.18
2-Amino-4-methylpyrimidine ≥98%
2-Amino-4-methylpyrimidine

Purity: 98% CAS No.: 108-52-1 Molecular Formula: C5H7N3 Formula Weight: 109.13 Melting Point: 158-161° Beilstein Reference: 108506

Formula: C₅H₇N₃
MW: 109.13 g/mol
Boiling Pt: 283
Melting Pt: 158…161 °C
Density: 1 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00006101
CAS Number: 108-52-1
EINECS: 203-591-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR