Tetramethylurea ≥99%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: N,N,N',N'-Tetramethylurea, 1,1,3,3-Tetramethylurea

A16295.18 A16295.30
A16295.18EA 37.2 EUR
A16295.18 A16295.30
Tetramethylurea ≥99%
Tetramethylurea

Purity: 99% CAS No.: 632-22-4 Molecular Formula: C5H12N2O Formula Weight: 116.16 Flash Point: 75°(167°F) Refractive Index: 1.4506 Odour: Mild Melting Point: -1° Boiling Point: 176-178° Solubility: Miscible with water Beilstein Reference: 773898 Merck Reference: 14,9230 MDL No.: MFCD00008319

Formula: C₅H₁₂N₂O
MW: 116.16 g/mol
Boiling Pt: 176…178 °C
Melting Pt: –1 °C
Density: 0.969
Flash Pt: 75 °C (167 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008319
CAS Number: 632-22-4
EINECS: 211-173-9
Merck Index: 12,09368

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR