1-(2,4-Difluorophenyl)-2-thiourea ≥97%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
N-(2,4-Difluorophenyl)thiourea, 1-(2,4-Difluorophenyl)thiourea, 2,4-Difluorophenylthiourea, (2,4-Difluorophenyl)thiourea
A16945.03
A16945.06
A16945.03EA
24.3
EUR
A16945.03
A16945.06
1-(2,4-Difluorophenyl)-2-thiourea ≥97%
1-(2,4-Difluorophenyl)-2-thiourea
Purity: 97%
CAS No.: 175277-76-6
Molecular Formula: C7H6F2N2S
Formula Weight: 188.20
Melting Point: 165-168°
UN No.: 2811
MDL No.: MFCD00041150
Formula:
(C₆H₃F₂)NC(S)NH₂ MW: 188,2 g/mol Boiling Pt: 272 °C (1013 hPa) Melting Pt: 165…168 °C Storage Temperature: Ambient |
MDL Number:
MFCD00041150 CAS Number: 175277-76-6 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...