Trichloromethyl chloroformate ≥98%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Chloroformic acid trichloromethyl ester, Diphosgene

A17444.09 A17444.18 A17444.30
A17444.09EA 51.8 EUR
A17444.09 A17444.18 A17444.30
Trichloromethyl chloroformate ≥98%
Trichloromethyl chloroformate

Purity: 98% CAS No.: 503-38-8 Molecular Formula: C2Cl4O2 Formula Weight: 197.83 Flash Point: 110°(230°F) Refractive Index: 1.4570 Odour: Pungent Melting Point: -57° Boiling Point: 128° Beilstein Reference: 970225 Merck Reference: 14,3334 UN No.: 3390 MDL No.: MFCD00015553

Formula: C₂Cl₄O₂
MW: 197.83 g/mol
Boiling Pt: 128 °C
Melting Pt: –57 °C
Density: 1.640
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00015553
CAS Number: 503-38-8
EINECS: 207-965-9
ADR: 6.1 I
Merck Index: 13,03368

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR