Benzyl bromoacetate ≥97%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: Benzyl 2-bromoacetate, Bromoacetic benzyl ester

A17630.0B A17630.18 A17630.30
A17630.0BEA 377 EUR
A17630.0B A17630.18 A17630.30
Benzyl bromoacetate ≥97%
Benzyl bromoacetate

Purity: 97% CAS No.: 5437-45-6 Molecular Formula: C6H5CH2CO2CH2Br Formula Weight: 229.08 Flash Point: 110°(230°F) Refractive Index: 1.5440 Odour: Pungent Boiling Point: 157-159°/10mm Beilstein Reference: 973658 MDL No.: MFCD00000190

Formula: C₉H₉BrO₂
Boiling Pt: 157…159 °C (10 mmHg)
Density: 1,460
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000190
CAS Number: 5437-45-6
EINECS: 226-611-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR