5-Chloroindole-2-carboxylic acid ≥98%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
5-Chloro-1H-indole-2-carboxylic acid
A18626.03
A18626.06
A18626.14
A18626.03EA
62.7
EUR
A18626.03
A18626.06
A18626.14
5-Chloroindole-2-carboxylic acid ≥98%
5-Chloroindole-2-carboxylic acid
Purity: 98%
CAS No.: 10517-21-2
Molecular Formula: C9H6ClNO2
Formula Weight: 195.60
Melting Point: 287° dec.
Beilstein Reference: 153229
Formula:
C₉H₆ClNO₂ MW: 195,6 g/mol Melting Pt: 287 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00005613 CAS Number: 10517-21-2 EINECS: 234-050-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...