4-Chlorobutyronitrile ≥97%
Supplier: Thermo Scientific
|
Danger
|
A18942.0B
A18942.18
A18942.30
A18942.0BEA
514
EUR
A18942.0B
A18942.18
A18942.30
4-Chlorobutyronitrile ≥97%
4-Chlorobutyronitrile
Purity: 97%
CAS No.: 628-20-6
Molecular Formula: C4H6ClN
Formula Weight: 103.55
Flash Point: 85°(185°F)
Refractive Index: 1.4440
Boiling Point: 195-197°
Beilstein Reference: 1633582
UN No.: 3276
MDL No.: MFCD00001972
Formula:
C₄H₆ClN MW: 103,55 g/mol Boiling Pt: 195…197 °C Melting Pt: –44 °C Density: 1,084 Flash Pt: 85 °C (185 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001972 CAS Number: 628-20-6 EINECS: 211-031-6 UN: 3276 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...