3-Amino-5-tert-butylisoxazole 97%

Supplier: Thermo Fisher Scientific
Warning

B20567.06 B20567.14 B20567.22
B20567.06EA 53.9 EUR
B20567.06 B20567.14 B20567.22
3-Amino-5-tert-butylisoxazole 97%
3-Amino-5-tert-butylisoxazole

Purity: 97% CAS No.: 55809-36-4 Molecular Formula: C7H12N2O Formula Weight: 140.19 Melting Point: 110-113° Beilstein Reference: 1100990

Formula: C₇H₁₂N₂O
Melting Pt: 110…113 °C
Storage Temperature: Ambient
MDL Number: MFCD00055620
CAS Number: 55809-36-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR