Sodium dichloroacetate 97
Supplier: Thermo Fisher Scientific
|
Warning
|
Synonyms:
Sodium 2,2-dichloroacetate
B21827.09
B21827.18
B21827.30
B21827.09EA
77.6
EUR
B21827.09
B21827.18
B21827.30
Sodium dichloroacetate 97
Sodium dichloroacetate
Purity: 97%
CAS No.: 2156-56-1
Molecular Formula: Cl2CHCO2Na
Formula Weight: 150.93
Melting Point: 197-199° dec.
MDL No.: MFCD00070489
Formula:
C₂HCl₂NaO₂ MW: 150.92 g/mol Melting Pt: 197…199 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00070489 CAS Number: 2156-56-1 EINECS: 218-461-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...