3-(Methylthio)propyl isothiocyanate ≥98%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
3-(Methylthio)propylisothiocyanate
B22150.09
B22150.18
B22150.09EA
58.7
EUR
B22150.09
B22150.18
3-(Methylthio)propyl isothiocyanate ≥98%
3-(Methylthio)propyl isothiocyanate
Purity: 98%
CAS No.: 505-79-3
Molecular Formula: CH3S(CH2)3NCS
Formula Weight: 147.25
Flash Point: 110°(230°F)
Refractive Index: 1.564
Odour: Pungent
Boiling Point: 254°
UN No.: 2922
MDL No.: MFCD00209521
Formula:
C₅H₉NS₂ MW: 147,27 g/mol Boiling Pt: 254 °C Density: 1,113 Flash Pt: 110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00209521 CAS Number: 505-79-3 EINECS: 208-020-3 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...