4-Amino-2,6-dimethylpyrimidine 99%

Supplier: Thermo Fisher Scientific
Warning

B22175.09 B22175.88
B22175.09EA 77.5 EUR
B22175.09 B22175.88
4-Amino-2,6-dimethylpyrimidine 99%
4-Amino-2,6-dimethylpyrimidine

Purity: 99% CAS No.: 461-98-3 Molecular Formula: C6H9N3 Formula Weight: 123.16 Melting Point: 184-186° Merck Reference: 14,5326 MDL No.: MFCD00006106

Formula: C₆H₉N₃
Boiling Pt: 250…254 °C (1013 hPa)
Melting Pt: 184…186 °C
Storage Temperature: Ambient
MDL Number: MFCD00006106
CAS Number: 461-98-3
EINECS: 207-320-1
Merck Index: 14,5326

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR