4-Propoxyphenol ≥98%
Supplier: Thermo Fisher Scientific
|
Warning
|
B22266.14
B22266.22
B22266.14EA
197
EUR
B22266.14
B22266.22
4-Propoxyphenol ≥98%
4-Propoxyphenol
Purity: 98%
CAS No.: 18979-50-5
Molecular Formula: C9H12O2
Formula Weight: 152.19
Flash Point:
Formula:
C₉H₁₂O₂ Boiling Pt: 249 °C (1013 hPa) Melting Pt: 55…58 °C Flash Pt: >110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002335 CAS Number: 18979-50-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...