Benzyltrimethylammonium iodide ≥98%
Supplier: Thermo Fisher Scientific
Synonyms:
N,N,N-Trimethyl-1-phenylmethanaminium iodide
B23520.06
B23520.22
B23520.06EA
0
EUR
B23520.06
B23520.22
Benzyltrimethylammonium iodide ≥98%
Benzyltrimethylammonium iodide
Purity: 98%
CAS No.: 4525-46-6
Molecular Formula: C6H5CH2N(CH3)3I
Formula Weight: 277.15
Melting Point: 177-182°
Beilstein Reference: 3917007
Formula:
C₁₀H₁₆IN MW: 277.15 g/mol Melting Pt: 179 |
MDL Number:
MFCD00051859 CAS Number: 4525-46-6 EINECS: 224-861-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...