3-Bromofuran ≥97% stabilised
Supplier: Thermo Scientific
|
Danger
|
Stabilisation:
Stabilised with calcium carbonate 0,5 %
B23543.06
B23543.03
B23543.14
B23543.06EA
167
EUR
B23543.06
B23543.03
B23543.14
3-Bromofuran ≥97% stabilised
3-Bromofuran
Stabilized with 0.5% calcium carbonate
Purity: 97%
CAS No.: 22037-28-1
Molecular Formula: C4H3BrO
Formula Weight: 146.98
Storage Temperature: -30°C to -10°C
Flash Point: 3°(37°F)
Refractive Index: 1.4960
Boiling Point: 102-104°
Beilstein Reference: 105337
UN No.: 1993
MDL No.: MFCD00005347
Formula:
C₄H₃BrO MW: 146,97 g/mol Boiling Pt: 102…104 °C Density: 1,635 Flash Pt: 3 °C (37 °F) Storage Temperature: Freezer |
MDL Number:
MFCD00005347 CAS Number: 22037-28-1 EINECS: 244-744-6 UN: 1993 ADR: 3,II |
Learn more
![](/m-ie.vwr.com/en_IE/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...