1-Adamantanemethanol ≥98%
Supplier: Thermo Fisher Scientific
B23831.06
B23831.14
B23831.06EA
63.4
EUR
B23831.06
B23831.14
1-Adamantanemethanol ≥98%
1-Adamantanemethanol
Purity: 98%
CAS No.: 770-71-8
Molecular Formula: C11H18O
Formula Weight: 166.26
Melting Point: 115-118°
Beilstein Reference: 1853339
Formula:
C₁₁H₁₈O Boiling Pt: 253 °C (1013 hPa) Melting Pt: 115…118 °C Storage Temperature: Ambient |
MDL Number:
MFCD00074751 CAS Number: 770-71-8 EINECS: 212-225-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...