Isopropyl salicylate ≥99%

Supplier: Thermo Fisher Scientific

Synonyms: Isopropyl 2-hydroxybenzoate

B24471.14 B24471.22
B24471.14EA 58.7 EUR
B24471.14 B24471.22
Isopropyl salicylate ≥99%
Isopropyl salicylate

Purity: 99% CAS No.: 607-85-2 Molecular Formula: C10H12O3 Formula Weight: 180.20 Flash Point: 93°(199°F) Boiling Point: 122°/18mm MDL No.: MFCD00035703

Formula: C₁₀H₁₂O₃
MW: 180.2 g/mol
Boiling Pt: 122 °C (18 mmHg)
Density: 1.09
Flash Pt: >93 °C (199 °F)
Storage Temperature: Ambient
MDL Number: MFCD00035703
CAS Number: 607-85-2
EINECS: 210-143-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR