5-Benzyloxyindole ≥94% may contain up to ca 7% toluene

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 5-Benzyloxy-1H-indole, 5-(Benzyloxy)-1H-indole

B25143.03 B25143.06 B25143.14
B25143.03EA 36.7 EUR
B25143.03 B25143.06 B25143.14
5-Benzyloxyindole ≥94% may contain up to ca 7% toluene
5-Benzyloxyindole

Purity: 94% CAS No.: 1215-59-4 Molecular Formula: C15H13NO Formula Weight: 223.28 Melting Point: 96-101° Beilstein Reference: 173532

Formula: C₁₅H₁₃NO
MW: 223.27 g/mol
Boiling Pt: 165 °C (0.1 mmHg)
Melting Pt: 96…101 °C
Storage Temperature: Ambient
MDL Number: MFCD00005676
CAS Number: 1215-59-4
EINECS: 214-930-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR