4-Bromo-m-toluonitrile ≥98%
Supplier: Thermo Scientific
|
Danger
|
B25377.03
B25377.06
B25377.14
B25377.03EA
36.3
EUR
B25377.03
B25377.06
B25377.14
4-Bromo-m-toluonitrile ≥98%
4-Bromo-m-toluonitrile
Purity: 98%
CAS No.: 41963-20-6
Molecular Formula: C8H6BrN
Formula Weight: 196.05
Flash Point: 96°(205°F)
Melting Point: 52-54°
Boiling Point: 129-130°/5mm
Beilstein Reference: 2354972
UN No.: 3439
MDL No.: MFCD00031538
Formula:
C₈H₆BrN MW: 196,05 g/mol Boiling Pt: 129…130 °C (5 mmHg) Melting Pt: 52…54 °C Flash Pt: 96 °C (205 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00031538 CAS Number: 41963-20-6 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...