3,5-Dibromobenzaldehyde ≥98%
Supplier: Thermo Fisher Scientific
|
Warning
|
B25495.03
B25495.06
B25495.14
B25495.03EA
0
EUR
B25495.03
B25495.06
B25495.14
3,5-Dibromobenzaldehyde ≥98%
3,5-Dibromobenzaldehyde
Purity: 98%
CAS No.: 56990-02-4
Molecular Formula: C7H4Br2O
Formula Weight: 263.93
Melting Point: 89-93°
Beilstein Reference: 2573432
Formula:
C₇H₄Br₂O MW: 263.92 g/mol Melting Pt: 89…93 °C Storage Temperature: Ambient |
MDL Number:
MFCD00156887 CAS Number: 56990-02-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...