5-Fluoropyridine-2-carbonitrile ≥98%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
5-Fluoro-2-pyridinecarbonitrile, 2-Cyano-5-fluoropyridine
H25956.06
H25956.03
H25956.06EA
163
EUR
H25956.06
H25956.03
5-Fluoropyridine-2-carbonitrile ≥98%
5-Fluoropyridine-2-carbonitrile
Purity: 98+%
CAS No.: 327056-62-2
Molecular Formula: C6H3FN2
Formula Weight: 122.10
Melting Point: 41-44°
MDL No.: MFCD08741371
Formula:
C₆H₃FN₂ MW: 122,1 g/mol Melting Pt: 41…44 °C Storage Temperature: Ambient |
MDL Number:
MFCD08741371 CAS Number: 327056-62-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...